Learn More
Abcam Streptavidin Protein, Native, Lyophilized, 1 mg
Description
- Protein Name: Streptavidin
- Source: Streptomyces avidinii
- Expression System: Native
- Applications: Flow Cyt, WB
- Form: Lyophilized
- Purification: Size-exclusion chromatography
- Formulation: Phosphate buffer, 0.58% sodium chloride, 0.013% sodium azide, pH 7.4
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Streptavidin |
| Substrate Properties | Protein-based Substrate |
| Shipping Condition | Blue Ice |
| Label or Dye | PE |
| Form | Lyophilized |
| Buffer | Phosphate buffer, 0.58% sodium chloride, 0.013% sodium azide, pH7.4 |
| Content And Storage | Store at 4°C. |
| Detection Location | In-Blot Detection |
| Formulation | Lyophilized from Phosphate Buffer, 0.58% Sodium chloride, 0.013% Sodium azide, pH 7.4 |
| Quantity | 1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.