Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Streptavidin Protein, Native, Lyophilized, Flow Cyt, 1 mg

Catalog No. 502707018
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7018 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7018 Supplier Abcam Supplier No. AB1331431MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Native Streptomyces avidinii streptavidin protein suitable for flow cytometry applications.

  • Protein Name: Streptavidin
  • Expression System: Native
  • Tested Applications: Flow Cyt
  • Form: Lyophilized
  • Purification Technique: Size-exclusion chromatography
  • Form: Lyophilized, pH 7.4. Contains 0.05% Sodium azide, Phosphate Buffer, 6.85% Sucrose, 0.58% Sodium chloride, and 0.1% BSA

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Streptavidin
Shipping Condition Blue Ice
Label or Dye Unconjugated
Content And Storage Store at 4°C.
Buffer Phosphate buffer, 6.85% sucrose, 0.58% sodium chloride, 0.1% BSA, 0.05% sodium azide, pH7.4
Form Lyophilized
Formulation Lyophilized from phosphate buffer, 6.85% sucrose, 0.58% sodium chloride, 0.1% BSA, 0.05% sodium azide, pH 7.4
Quantity 1 mg
For Use With (Application) Flow Cytometry
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.