Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Streptavidin Protein, Native, SureLight APC, 1 mg
Description
- Protein Name: Streptavidin
- Source: Streptomyces avidinii
- Expression System: Native
- Tested Applications: Flow Cyt, FRET
- Purification: Size-exclusion chromatography
- Form: Lyophilized; includes 0.01% Sodium azide, 0.88% Sodium chloride, 0.121% Tris, 0.1% BSA, pH 8.2
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Biotin |
| Substrate Properties | Protein-based Substrate |
| Buffer | 0.88% sodium chloride, 0.121% Tris, 0.1% BSA, 0.01% sodium azide, pH8.2 |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Label or Dye | APC |
| Shipping Condition | Blue Ice |
| Formulation | Lyophilized, pH 8.2, 0.01% sodium azide, 0.88% sodium chloride, 0.121% Tris, 0.1% BSA |
| Quantity | 1 mg |
| For Use With (Application) | Flow Cytometry, FRET |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?