Learn More
Abcam Streptavidin Protein, Streptomyces Avidinii, Native, 1 MG
Description
- Protein Name: Streptavidin
- Species: Streptomyces avidinii
- Expression System: Native
- Applications: Flow Cyt, FRET
- Form: Lyophilized
- Purification Technique: Size-exclusion chromatography
- Formulation: pH 8.2; contains 0.01% Sodium azide, 0.88% Sodium chloride, 0.121% Tris, and 0.1% BSA
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Protein-based Substrate |
| Target Molecule | Biotin |
| Buffer | 0.121% Tris, 0.88% sodium chloride, 0.1% BSA, 0.01% sodium azide, pH8.2 |
| Label or Dye | APC |
| Content And Storage | Store at -20°C. |
| Form | Lyophilized |
| Shipping Condition | Blue Ice |
| Formulation | Lyophilized, pH 8.2, 0.01% Sodium azide, 0.88% Sodium chloride, 0.121% Tris, 0.1% BSA |
| Quantity | 1 mg |
| For Use With (Application) | Flow Cytometry, FRET |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.