Learn More
AdipoGen Streptavidin R-PE
SDP
Supplier:  AdipoGen AG35B0014RC050
Encompass_Preferred
Recombinant protein. Recombinant streptavidin from Streptomyces avidinii. Binds to any biotinylated reagents. Endotoxin . Liquid. Contains PBS and 0.1% sodium azide. Streptavidin is a non-glycosylated protein isolated from Streptomyces avidinii. It forms a homotetramer and strong non-covalent specific complex with biotin one molecule of biotin per subunit of streptavidin. Streptavidin R-PE is typically used for flow cytometry, microarrays, ELISA and other applications that require either high sensitivity or simultaneous multicolor detection. Phycoerythrin is a member of a family of proteins called phycobiliproteins, which are derived from cyanobacteria and eukaryotic algae and exhibit extremely bright fluorescence and high quantum yields.
Catalog No. 50-256-3076
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.