Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus A Rabbit anti-Streptococcus, Unconjugated, Polyclonal, MilliporeSigma™

Catalog No. ABX850210MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABX850210MI 10 mg
ABX85021KMI 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABX850210MI Supplier MilliporeSigma Supplier No. ABX850210KC
Add to Cart
Add to Cart
Request a Quote

Rabbit Polyclonal Antibody

Streptococcus A Polyclonal antibody specifically detects Streptococcus A in Streptococcus samples. It is validated for Enzyme Immunoassay (ELISA), Immunodiffusion.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Streptococcus A (10mg)
Applications ELISA, Immunodiffusion
Classification Polyclonal
Conjugate Unconjugated
Gene Alias Rabbit anti-Strptococcus A
Host Species Rabbit
Immunogen Fragment of Streptococcus A after disruption
Purification Method Ion-Exchange Purified
Quantity 10 mg
Research Discipline Infectious Diseases
Primary or Secondary Primary
Test Specificity Specific for Group A Streptococcus
Target Species Streptococcus species
Content And Storage 2 to 8°C. For long-term storage, freeze at -20°C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.