Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptococcus agalactiae Monoclonal Antibody (Strep12-424.1), Invitrogen™
GREENER_CHOICE

Mouse Monoclonal Antibody

$472.00 - $1160.00

Specifications

Antigen Streptococcus agalactiae
Clone Strep12-424.1
Concentration 0.75 mg/mL
Applications ELISA
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIMA546154
View Documents
Invitrogen™
MA546154
100 μg
Each for $472.00
Only null left
Add to Cart
 
01-671-880
GREENER_CHOICE
View Documents
Invitrogen™
MA110871
500 μg
Each for $632.00
Only null left
Add to Cart
 
PIMA546181
View Documents
Invitrogen™
MA546181
1 mg
Each for $1,160.00
Only null left
Add to Cart
 
Description

Description

MA1-10871 specifically detects Streptococcus agalactiae. This antibody does not cross react with the Streptococcus species: dysgalactiae, uberis, bovis, and faecalis; the Mycoplasma species: bovis, californicum, and genitalium; as well as Staphlyococcus aureus, E. coli, and Corynebacterium bovis. MA1-10871 has been successfully used in ELISA applications. The MA1-10871 immunogen is UV-inactivated Strep agalactiae cells isolated from mastitic milk.

Once thought to infect only cows, where it produces mastitis, Streptococcus agalactiae is now known to be able to cause serious disease, bacteremia and meningitis, in immunocompromised individuals and in neonates. Infections in the newborn are the most common and are extremely serious. They are of two types. Early onset of infection, usually occuring within 5 days of birth, is manifested by bacteremia and pneumonia. This infection is contracted vertically, as the baby passes through the birth canal. The other type of infection is a meningitis that occurs between the 10th and 60th days of life. The origin of this infection is not known, but it is not believed to be the mother.
Specifications

Specifications

Streptococcus agalactiae
0.75 mg/mL
Monoclonal
Liquid
RUO
PBS with no preservative; pH 7.4
UV-inactivated Strep agalactiae cells isolated from mastitic milk.
Primary
-20°C, Avoid Freeze/Thaw Cycles
Strep12-424.1
ELISA
Unconjugated
Mouse
Bacteria
s agalactiae; S. agalactiae
IgM
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.