Learn More
Invitrogen™ Streptococcus Group A Recombinant Rabbit Monoclonal Antibody (0435)

Description
No cross reactivity with: Streptococci groups B, C, D, F & G + other bacteria.
Specifications
Specifications
| Antigen | Streptococcus Group A |
| Applications | Lateral Flow |
| Classification | Recombinant Monoclonal |
| Clone | 0435 |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.5 |
| Host Species | Rabbit |
| Purification Method | Protein A |
| Quantity | 1 mL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.