Learn More
Invitrogen™ Streptococcus Group A Monoclonal Antibody (HSA12-336.1), Invitrogen™

Description
MA1-10700 detects Streptococci Group A. MA1-10700 has been successfully used in ELISA applications. The MA1-10700 immunogen is UV-inactivated Group A Streptococcus cells.
Specifications
Specifications
| Antigen | Streptococcus Group A |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | HSA12-336.1 |
| Concentration | 5.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Host Species | Mouse |
| Immunogen | UV-inactivated Group A Streptococcus cells. |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.