Learn More
Invitrogen™ Streptococcus Group B Monoclonal Antibody (HSB11-209.5), Invitrogen™
Shop All Thermo Scientific Products

Description
MA1-10703 detects Streptococci Group B. MA1-10703 has been successfully used in ELISA applications. The MA1-10703 immunogen is UV-inactivated Group B Streptococcus cells, types Ia, Ib, Ic, II, and III.
Specifications
Specifications
| Antigen | Streptococcus Group B |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | HSB11-209.5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Host Species | Mouse |
| Immunogen | UV-inactivated Group B Streptococcus cells, types Ia, Ib, Ic, II, and III. |
| Purification Method | Protein G |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.