Learn More
Invitrogen™ Streptococcus Group B Monoclonal Antibody (HSB12-396.1)

Description
MA1-10706 detects Streptococci Group B. MA1-10706 has been successfully used in ELISA applications. The MA1-10706 immunogen is UV-inactivated Group B Streptococcus cells, types Ia, Ib, Ic, II, and III.
Specifications
Specifications
| Antigen | Streptococcus Group B |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | HSB12-396.1 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Host Species | Mouse |
| Immunogen | UV-inactivated Group B Streptococcus cells, types Ia, Ib, Ic, II, and III. |
| Purification Method | Protein G |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.