Learn More
Invitrogen™ Streptococcus pneumoniae Monoclonal Antibody (XI126)

Description
MA1-10834 detects Streptococcus pneumoniae. MA1-10834 has been successfully used in ELISA and Western blot applications. The MA1-10834 immunogen is Heat-killed S. pneumoniae strain R36A.
Specifications
Specifications
| Antigen | Streptococcus pneumoniae |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | XI126 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene Alias | Pneumococcus; s pneumoniae; S. pneumoniae |
| Host Species | Mouse |
| Immunogen | Heat-killed S. pneumoniae strain R36A. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.