Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Streptomycin ELISA Kit

Catalog No. 89335458 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-458 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-458 Supplier Abnova™ Supplier No. KA5346

Competitive enzyme-linked immunosorbent assay

Streptomycin ELISA Kit is a competitive enzyme-linked immunosorbent assay for the quantitative measurement of Streptomycin in tissue, honey and milk.

Specifications

For Use With (Application) Quantitative
Quantity 1 Kit
Regulatory Status RUO
Storage Requirements Store the kit at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.