Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streptozotocin, MP Biomedicals™

Catalog No. MP21005575 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP21005575 500 mg
MP21005576 50 mg
MP21005571 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP21005575 Supplier MP Biomedicals, Inc Supplier No. 02100557.5
Add to Cart
Add to Cart

Used in the treatment of pancreatic islet cell tumors. Streptozotocin, MP Biomedicals is a glucosamine-nitrosourea, a DNA-alkylating agent that enters cells via the GLUT2 glucose transport protein. It is used to treat malignant insulinoma and to induce Ty

Streptozocin is used to treat pancreatic islet cell tumors, including malignant insulinoma. It is also used to induce Type 1 diabetes in animals for medical research purposes.

  • An N-nitroso-containing compound that acts as a nitric oxide donor in pancreatic islets
  • Induces death of insulin-secreting cells, producing an animal model of diabetes
  • Potent DNA methylating agent that induces chromosomal breakage
  • Cytotoxic to neuroendocrine tumor cell lines that express the GLUT2 glucose transporter
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18883-66-4
Molecular Formula C8H15N3O7
Molecular Weight (g/mol) 265.222
InChI Key ZSJLQEPLLKMAKR-UHEPRFFZSA-N
Synonym N-(Methylnitrosocarbamoyl)-α-D-glucosamine, Streptozocin, STZ, 2-Deoxy-2-(3-methyl-3-nitrosoureido)-D-glucopyranose
PubChem CID 45357367
IUPAC Name 1-methyl-1-nitroso-3-[(2R,4R,5S,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]urea
SMILES CN(C(=O)NC1C(C(C(OC1O)CO)O)O)N=O

Specifications

Physical Form Powder
Melting Point 121°C (decomposition) (literature)
Extinction Coefficient EmM (228nm): 6.36 (ethanol)
Quantity 500 mg
Solubility Information Soluble in water (50mg/mL, clear to slightly hazy, light yellow solution) and ethanol.
Formula Weight 265.22
Percent Purity ∼75% α-anomer basis, ≥97%
Recommended Storage −20°C, desiccate, store under nitrogen
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.