Learn More
Description
- Prepacked with StrepTactin Sepharose High Performance
- Binding capacity Approx. 6 mg Strep II-tagged protein/ml medium
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
- Generated with 0.5M NaOH
- 1 and 5mL columns
Media
- Ligand: Streptactin
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: >7
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
Specifications
| Product Type | StrepTactin Sepharose HP prepacked column |
| Bed Volume | 1mL |
| Diameter (English) Inner | 0.62 in. |
| Diameter (Metric) Inner | 16mm |
| Quantity | 1 x 5mL |
| Height (English) | 0.98 in. |
| Height (Metric) | 25mm |
| Flow Rate | <20 mL/min. |
| Material | Polypropylene |
| Ligand Type | StrepTactin (∼5mg/mL medium) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.