Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva StrepTrap XT Chromatography Column

Catalog No. 09920073 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
1 x 1 mL
1 x 5 mL
5 x 1 mL
5 x 5 mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-920-073 5 x 5 mL
09-920-070 1 x 1 mL
09-920-071 5 x 1 mL
09-920-072 1 x 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 09-920-073 Supplier Cytiva Supplier No. 29401323
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

StrepTrap XP is a column packed with Strep-Tactin™ XT Sepharose resin to purify Strep-tag™ II and Twin-Strep-tag™ recombinant proteins.

  • For use with Strep-Tactin™ XT Sepharose™
  • Prepacked in chromatography columns for convenience
TRUSTED_SUSTAINABILITY

Specifications

Bed Volume 5 mL
Diameter (Metric) Inner 16 mm
Quantity 5 x 5 mL
Height (Metric) 25 mm
Flow Rate 5 mL/min.
Column Type Affinity Column
For Use With (Application) Purification of recombinant proteins tagged with Strep-tag™ II and Twin-Strep-tag™
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.