Learn More
Grainger Stretch Wrap Cast Standard Duty
Supplier:  Grainger 15A993
Encompass_Preferred
The Grainger 15A993 Stretch Wrap is a 70 gauge, hand wrap stretch film that provides load containment and product protection. It is constructed from a linear low-density polyethylene and is recommended for light-duty wrapping, bundling, and palletizing applications. This stretch wrap film measures 20 inches wide x 1500 feet long, provides 130% prestretch, and has a break strength of 1550 PSI. The 70 gauge thickness offers good puncture resistance. It can be used with most standard hand wrap dispensers and is an economical choice for light-duty stretch wrapping needs.
Catalog No. 50-275-1709
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.