Learn More
Grainger Stretch Wrap Cast Standard Duty
Supplier:  Grainger 15A988
Encompass_Preferred
The Grainger 15A988 Stretch Wrap is a 70 gauge, hand wrap stretch film that provides load containment and product protection. It is constructed from a linear low-density polyethylene and is recommended for light-duty wrapping, bundling, and palletizing applications. This 12" x 1500' clear stretch wrap film features excellent cling to adhere to itself and the load. It can stretch up to 200% to ensure a tight wrap and has 55% memory to retain tension. The film resists punctures and tears during application and transportation. This cost-effective stretch wrap is an ideal choice for light-duty packaging and unitizing needs.
Catalog No. 50-275-3340
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.