Learn More
Ampco Safety Tools Striking Open Wrench (Metric)
SDP
Catalog No. 19175089 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Drive Size:
14 mm
17 mm
19 mm
22 mm
24 mm
25 mm
26 mm
27 mm
28 mm
29 mm
30 mm
32 mm
33 mm
35 mm
36 mm
37 mm
38 mm
41 mm
42 mm
46 mm
47 mm
50 mm
52 mm
54 mm
55 mm
56 mm
60 mm
63 mm
65 mm
67 mm
70 mm
71 mm
75 mm
77 mm
80 mm
82 mm
85 mm
88 mm
90 mm
92 mm
94 mm
95 mm
100 mm
105 mm
110 mm
115 mm
120 mm
125 mm
48 product options available for selection
Product selection table with 48 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Drive Size
19-175-089 55 mm
19-175-005 100 mm
19-175-006 105 mm
19-175-012 110 mm
19-175-015 115 mm
19-175-017 120 mm
19-175-018 125 mm
19-175-023 14 mm
19-175-027 17 mm
19-175-030 19 mm
19-175-040 22 mm
19-175-044 24 mm
19-175-045 25 mm
19-175-048 26 mm
19-175-049 27 mm
19-175-052 28 mm
19-175-053 29 mm
19-175-057 30 mm
19-175-061 32 mm
19-175-062 33 mm
19-175-066 35 mm
19-175-068 36 mm
19-175-069 37 mm
19-175-071 38 mm
19-175-073 41 mm
19-175-077 42 mm
19-175-081 46 mm
19-175-082 47 mm
19-175-086 50 mm
19-175-087 52 mm
19-175-088 54 mm
19-175-090 56 mm
19-175-092 60 mm
19-175-093 63 mm
19-175-094 65 mm
19-175-095 67 mm
19-175-097 70 mm
19-175-098 71 mm
19-175-099 75 mm
19-175-100 77 mm
19-175-101 80 mm
19-175-102 82 mm
19-175-103 85 mm
19-175-104 88 mm
19-175-107 90 mm
19-175-112 92 mm
19-175-113 94 mm
19-175-114 95 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 48 options available.
48 options
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
Catalog No. 19-175-089 Supplier Ampco Safety Tools Supplier No. WSO55
Add to Cart
Add to Cart
AMPCO and other copper-based tools should not be used in direct contact with acetylene, ethylene oxide and some explosives such as amatol, DBX and lead azide.
AMPCO™ Open Striking Wrench performs in high-risk and demanding environments.
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Warranty and Services
Limited Lifetime Warranty
Specifications
Product Type Striking Open Wrench
Drive Size 55 mm
Material Aluminum Bronze
Certifications/Compliance Factory Mutual (FM) Approved
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.