Learn More
Boston Bioproducts Inc Stripping Buffer (4X, Mild) - 250ml
Supplier:  Boston Bioproducts Inc BP97250ML
Encompass_Preferred
The 4X Stripping Buffer (Mild) containing 200 mM Glycine, is used for breaking antibody-antigen interactions, resulting in removal of primary and secondary antibodies from their target proteins on probed Western blot membranes. The use of this buffer minimizes protein denaturation, preserves epitope integrity, and is suitable for re-probing, saving time without the need for running multiple gels and performing transfers.
Catalog No. NC9498788
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.