Learn More
Cytiva Petri Dishes without Absorbant Pads, 50 mm, Gamma Irradiated (100/pkg)
Catalog No. NC0874606 Shop All Cytiva Products
Encompass
Unique patented design for easy handling and storage-Easy to use. Opens easily with one hand, yet closes to a tight seal. -Uses less space on the lab bench or in the incubator with easy stacking base.-Gamma irradiated for sterilization. No dangerous EtO residuals.-Available with or without absorbent pads.-Bulk packaging for additional value. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0874606 Supplier Cytiva Supplier No. 7242
Unique patented design for easy handling and storage-Easy to use. Opens easily with one hand, yet closes to a tight seal. -Uses less space on the lab bench or in the incubator with easy stacking base.-Gamma irradiated for sterilization. No dangerous EtO residuals.-Available with or without absorbent pads.-Bulk packaging for additional value. This product is not approved for use in medical, clinical, surgical or other patient applications.