Learn More
Med Vet International Sterile Water For Injection, Diluent, 250mL, Each
Supplier:  Med Vet International RXWATER250INJV
Encompass_Preferred
Sterile water for injection is suitable for use as a diluent for the preparation of pharmaceutical solutions. Single-dose vial contains no preservatives. Not suitable for IV injection without first having been made isotonic by addition of suitable solutes.
Catalog No. 50-282-8421
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.