Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STRN1/STRN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100865
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100865 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100865 Supplier Invitrogen™ Supplier No. PA5100865
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total STRN1/3.

Binds calmodulin in a calcium dependent manner. May function as scaffolding or signaling protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STRN1/STRN3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene STRN
Gene Accession No. O43815, O55106, P58405, P70483, Q13033, Q9ERG2
Gene Alias AU022939; Cell cycle autoantigen SG2NA; cell cycle S/G2 nuclear autoantigen; cell-cycle autoantigen SG2NA; D130055P19; Gs2na; nuclear autoantigen; PPP2R6A; PPP2R6B; protein phosphatase 2 regulatory subunit B'''alpha; protein phosphatase 2 regulatory subunit B'''beta; s/G2 antigen; Sg2na; SG2NA beta isoform; striatin; striatin 3; striatin, calmodulin binding protein; striatin, calmodulin binding protein 3; striatin, calmodulin-binding protein; striatin-3; striatin-3 35 kDa; striatin-3-gamma; Strn; STRN3
Gene Symbols STRN, STRN3
Host Species Rabbit
Immunogen A synthesized peptide derived from human STRN3(Accession Q13033), corresponding to amino acid residues F305-Y355.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114520, 268980, 29149, 29966, 6801, 94186
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.