Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strong Ammonia Solution, NF, 27-31%, Spectrum™ Chemical
SDP

Catalog No. 18607032 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
2.5 L
4 L
6 x 500 mL
20 L
4 x 2.5 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
Poly-Coated Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-032 200 L Drum
18-607-027 2.5 L Poly-Coated Glass Bottle
18-607-026 6 x 500 mL Amber Glass Bottle
18-607-028 4 L Poly-Coated Glass Bottle
18-607-029 4 x 2.5 L Poly-Coated Glass Bottle
18-607-030 4 x 4 L Poly-Coated Glass Bottle
18-607-031 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-032 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AM180200LTBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

AM180, 1336-21-6, NH3(aq)

Spectrum™ Chemical Strong Ammonia Solution, NF has many uses such as in topical pharmaceutical solutions destined for treating insect bites. All Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
IUPAC Name amine hydrate
SMILES N.O

Specifications

CAS Max % 1
Quantity 200 L
Percent Purity 27 to 31%
Grade NF
Packaging Drum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.