Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Strong Iodine Solution, 5% (w/v), USP, Spectrum™ Chemical
SDP

Catalog No. 18607039 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
Drum
Poly Drum
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-039 200 L Drum
18-607-033 100 mL Amber Glass Bottle
18-607-034 500 mL Amber Glass Bottle
18-607-035 6 x 500 mL Amber Glass Bottle
18-607-036 4 L Amber Glass Bottle
18-607-037 4 x 4 L Amber Glass Bottle
18-607-038 20 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18-607-039 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. IO106200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

IO106

Spectrum™ Chemical Strong Iodine Solution, 5 Percent (w/v) (DEA List I Chemical), USP is used primarily as an antiseptic and disinfectant in pharmaceutical applications. All Spectrum Chemical USP products, including all USP iodine products, are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7553-56-2 , 7732-18-5
Molecular Formula I2
Molecular Weight (g/mol) 253.81
MDL Number MFCD00011355 MFCD00164163
InChI Key PNDPGZBMCMUPRI-UHFFFAOYSA-N
IUPAC Name diiodine
SMILES II

Specifications

CAS Max % 0.05, 0.95
Grade USP
Packaging Drum
Quantity 200 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.