Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Strontium Nitrate
SCI_ED

Catalog No. S25589A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Lab Grade
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25589A 500 g Lab Grade
S25589 100 g Lab Grade
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25589A

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10042-76-9
Molecular Formula N2O6Sr
Molecular Weight (g/mol) 211.63
MDL Number MFCD00011248
InChI Key DHEQXMRUPNDRPG-UHFFFAOYSA-N
Synonym strontium nitrate, strontium dinitrate, nitric acid, strontium salt, strontium ii nitrate 1:2, unii-bdg873aqzl, nitrate de strontium french, hsdb 787, strontium nitrate sr no3 2, bdg873aqzl, nitric acid, strontium salt 2:1
PubChem CID 24848
IUPAC Name strontium(2+) dinitrate
SMILES [Sr++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color White
Quantity 500 g
Assay Percent Range >95%
Linear Formula Sr(NO3)2
Formula Weight 211.63
Grade Laboratory
Packaging Lab Grade
Chemical Name or Material Strontium Nitrate
Safety handling Check Alert storage code Red
ShelfLife 5 years
DOTInformation DOT Class 5.1, III, : Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.