Learn More
Strem, An Ascensus Company CAS# 147658-82-0. 500mg. Bis(tri-isopropylcyclopentadienyl)strontium 98%.
SDP
Supplier:  Strem, An Ascensus Company 381200500MG
Encompass_Preferred
CAS# 147658-82-0. 500mg. Bis(tri-isopropylcyclopentadienyl)strontium 98%. . Molecular Weight: 470.29. Molecular Formula: [(C3H7)3C5H2]2Sr. Color/form: yellow liq. Strem# 38-1200. http://www.strem.com/catalog/v/38-1200/
Catalog No. 50-206-5538
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.