Learn More
Apexbio Technology LLC Triflurdine (Viroptic) 70-00-8 50mg
SDP
Supplier:  Apexbio Technology LLC A854250
Encompass_Preferred
Triflurdine Viroptic (70-00-8) is a small-molecule antiviral agent targeting herpes simplex virus (HSV) It is designed to incorporate into viral DNA during replication thereby interfering with nucleic acid synthesis and impairing viral proliferation Triflurdine Viroptic exerts its biological activity primarily through incorporation into viral DNA disrupting replication In in vitro studies Triflurdine Viroptic demonstrates antiviral activity with IC50 values ranging from approximately 1 0 to 10 0 M against HSV strains Based on these pharmacological properties Triflurdine Viroptic holds research potential in investigations of antiviral mechanisms viral resistance pathways and herpesvirus-host interactions
Catalog No. 50-000-95272
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.