Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Strontium sulfate, Puratronic™, 99.9965% (metals basis)

Manufacturer:  Alfa Aesar™ 1287014

Quantity 25g

 View more versions of this product

Catalog No. AA1287014


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

7759-02-6
183.68
14,8848
Puratronic™
SrSO4
MFCD00036274
Strontium sulfate

Specifications

7759-02-6
SrSO4
MFCD00036274
3.960g/mL
White
25g
99.9965%, (metals basis)
Powder
183.68
14,8848
Odorless
1600°C
Strontium sulfate
Puratronic™

Safety and Handling

SDS & Safety Documents

RTECSNumber : WT1210000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit