Learn More
Thermo Scientific™ Streptococcal Grouping Kit using Latex Agglutination
Catalog No. OXDR0592G
Change view
Click to view available options
Description:
Enzyme Extraction Reagent
Grouping Kit
Polyvalent Positive Control
Reagent A
Reagent B
Reagent C
Reagent D
Reagent F
Reagent G
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
OXDR0592G Polyvalent Positive Control
OXDR0585A Grouping Kit
OXDR0586G Reagent A
OXDR0587G Reagent B
OXDR0588G Reagent C
OXDR0589G Reagent D
OXDR0590G Reagent F
OXDR0591G Reagent G
OXDR0593G Enzyme Extraction Reagent
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Catalog No. OXDR0592G Supplier Thermo Scientific™ Supplier No. DR0592G
Add to Cart
Add to Cart
This item is not returnable. View return policy
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using this test kit.
Rapidly identify streptococcal groups A, B, C, D, F, and G by latex agglutination using Thermo Scientific™ Oxoid™ Streptococcal Grouping Kit. This rapid latex agglutination test is designed to deliver simple, rapid and sensitive results for the identification of streptococcal groups A, B, C, D, F, and G . The test utilizes latex particles coated with group-specific antibodies which will agglutinate in the presence of homologous antigens.
Streptococci are facultatively anaerobic, catalase-negative, Gram-positive cocci that occur as pairs or chains. They cause a wide variety of clinical diseases including pharyngitis, scarlet fever, otitis media, impetigo, meningitis, neonatal sepis, pneumonia, endocarditis, osteomyelitis, and necrotizing fasciitis.
Use this rapid, simple and sensitive test for the identification of streptococcal groups A, B, C, D, F, and G.
  • Rapid—results in two minutes
  • Simple—easy to implement
  • Visual—easy-to-read results
  • Sensitive—use of latex technology makes this test more sensitive
  • All reagents available separately
Lancefield showed that the majority of pathogenic streptococci possess specific carbohydrate antigens, which permit the classification of streptococci into groups. These streptococcal group antigens can be extracted from the cells and their presence demonstrated with latex particles previously coated with group-specific antibodies. These latex particles will agglutinate in the presence of homologous antigen, but will remain in smooth suspension in the absence of such antigen. The Oxoid Streptococcal Grouping Kit is such a latex agglutination test for the identification of the streptococcal groups and reagents are provided for groups A, B, C, D, F and G. The use of a enzymatic extraction procedure considerably shortens the time required for antigen extraction and much improves the antigen yield, partially for Group D streptococci.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Control Sets Positive
Description Polyvalent Positive Control
Type Positive Control
Material Polyvalent
Organism Group Group A, B, C, D, F and G
Quantity 50 Tests