Learn More
Description
PA1-015 detects human and mouse carboxyl terminus of hsc70-interacting protein (CHIP). PA1-015 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ~35 kDa protein representing CHIP from COS-1 cells overexpressing the human gene and a non-specific band at ~85 kDa. PA1-015 was used successfully in the western blot analysis of CHIP using human MCF7 cell lysates. The PA1-015 immunogen is a synthetic peptide corresponding to residues V(218) D E K R K K R D I P D Y L C(232) of human CHIP. This peptide (Cat. # PEP-179) is available for use in neutralization and control experiments.
Specifications
Specifications
| Antigen | STUB1 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | STUB1 |
| Gene Accession No. | Q9UNE7, Q9WUD1 |
| Gene Alias | 0610033N24Rik; 2210017D18Rik; 2310040B03Rik; Antigen NY-CO-7; AW046544; Carboxy terminus of Hsp70-interacting protein; CHIP; CLL-associated antigen KW-8; E3 ubiquitin-protein ligase CHIP; heat shock protein A binding protein 2 (c-terminal); HSPABP2; LA16c-313D11.6; NY-CO-7; PP1131; RING-type E3 ubiquitin transferase CHIP; SCAR16; SDCCAG7; serologically defined colon cancer antigen 7; STIP1 homology and U box-containing protein 1; STIP1 homology and U-box containing protein 1; STIP1 homology and U-box containing protein 1, E3 ubiquitin protein ligase; Stub1; UBOX1 |
| Gene Symbols | STUB1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
