Learn More
Eisco™ Student Basic Organic Chemistry Molecular Model Set
SCI_ED
Catalog No. S96541 Shop All Eisco Products
Change view
Click to view available options
Material:
Plastic
Catalog No. Material
S96541 Plastic
1 options
Catalog No. S96541 Supplier Eisco™ Supplier No. SETSNR
Add to Cart
Add to Cart
The Student Basic Organic Chemistry Molecular Model Set lets students visualize three-dimensional molecule structures and understand molecular formulas.
Hard plastic pails of different colors represent atoms; each has holes at correct angles to be connected with flexible plastic 'bonds. Medium links represent single covalent bonds while long links can be used for double and triple covalent bonds.
  • 12 Gray metal (2 holes)
  • 10 Gray metal (3 holes)
  • 10 Gray metal (6 holes)
  • 42 Red oxygen (2 holes)
  • 12 Red oxygen (4 holes)
  • 18 Red oxygen (1 holes)
  • 12 Yellow sulfur (4 holes)
  • 15 Blue nitrogen (3 holes)
  • 45 Black carbon (4 holes)
  • 12 Purple phosphorus (4 holes)
  • Six purple phosphorus (3 holes)
  • 36 White hydrogen (1 hole)
  • 170 Medium gray links
  • 100 Long gray links
Specifications
Includes Gray Metal, red Oxygen, yellow Sulfur, blue Nitrogen, black Carbon, purple Phosphorus, white Hydrogen and medium gray and long gray links
Material Plastic
Product Type Molecular Model, Student Basic Organic Chemistry Set