Learn More
Student-Grade Tuning Fork Sets
SCI_ED
Catalog No. p-2836507 Shop All Medivibe Technology Ltd. Products
Change view
Click to view available options
Quantity:
8/Set
Catalog No. Quantity
S68770 8/Set
1 options
This item is not returnable. View return policy
Catalog No. S68770 Supplier Medivibe Technology Ltd. Supplier No. 7008STEC
Only null left
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Economical set.
  • Eight-piece set includes forks with 256, 288, 320, 341.1, 384, 426.7, 480 and 512Hz
  • 13-Piece set includes forks with 261.6, 277.1, 293.7, 311.1, 329.6, 349.2, 369.9, 392, 415.30, 440, 466.1, 493.9 and 523.2Hz
Specifications
Product Type Tuning Fork Set
Includes Eight piece set includes forks with 256, 288, 320, 341.1, 384, 426.7, 480 and 512Hz
Quantity 8/Set