Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Student Gratings, 13,400 Lines/in.
SDPSCI_ED

Catalog No. S139474 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Grade:
5 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S139474 5 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S139474 Supplier United Scientific™ Supplier No. DFG525PK/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

Quality linear gratings protected by two glass plates.

  • Quality linear gratings protected by two glass plates, 60 mm x 45 mm
  • Pack of five quality linear gratings in slide viewers
  • Viewing area measures approximately 35 mm x 25 mm
  • Grating: 13,400 lines per inch

Specifications

Grade 5 to 12
Format Model
Languages English
Subject Physics

Warning: Cancer and reproductive harm

Prop 65

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.