Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STX3 Monoclonal Antibody (OTI5D10), TrueMAB™, OriGene
SDP

Catalog No. 501683810
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-3810 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-3810 Supplier OriGene Supplier No. TA811091
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

STX3 is otentially involved in docking of synaptic vesicles at presynaptic active zones.

Specifications

Antigen STX3
Applications Western Blot
Classification Monoclonal
Clone OTI5D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene STX3
Gene Accession No. Q08849, Q13277, Q64704
Gene Alias STX3A
Gene Symbols Stx3
Host Species Mouse
Immunogen Full length human recombit protein of human STX3 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20908, 6809, 81802
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.