Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STX7 Monoclonal Antibody (CL0257)
GREENER_CHOICE

Catalog No. PIMA531334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA531334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA531334 Supplier Invitrogen™ Supplier No. MA531334
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: GVGGDPAQLA QRISSNIQKI TQCSVEIQRT LNQLGTPQDS PELRQQLQQK QQYTNQLAKE TDKYIKEFGS LPTTPSEQRQ RKIQKDRLVA EFTTSLTNFQ KVQRQAAERE KEFVARVRAS SRVSGSFPED SSKERNLVSW ESQTQPQV Highest antigen sequence indentity to the following orthologs: Rat - 94%, Mouse - 97%.

ZCRB1 is a nuclear protein first identified in a differential display screen involving morphine-dependence-related genes. Its expression is increased following morphine treatment and has been observed to be elevated in HepG2 cells. ZCRB1 contains a CCHC-type zing finger RNA-binding motif and can interact with the DBA-binding domain of the stem cell regulator C/EBP transcription factors. ZCRB1 was identified as one of the protein components of U11/U12 snRNPs, which are components of U12-type spliceosome and function as a molecular bridge connecting both ends of the intron, suggesting ZCRB1 may play a key role in U12-type splicing.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STX7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Monoclonal
Clone CL0257
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Stx7
Gene Accession No. O15400
Gene Alias AI315064; AI317144; AW107239; STX7; Syn7; syntaxin 7; Syntaxin7; syntaxin-7; syntaxin-7-like; syntaxin-7-like protein; Unknown (protein for MGC:128117)
Gene Symbols Stx7
Host Species Mouse
Immunogen Recombinant protein fragment of Human STX7 (Product #RP-95935)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8417
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.