Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Styrene-4-sulfonic acid sodium salt

Catalog No. p-7051636
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4169018 50 g
AA4169030 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4169018 Supplier Thermo Scientific Chemicals Supplier No. 04169018
Only null left
Add to Cart

CAS: 2695-37-6 | C8H7NaO3S | 206.19 g/mol

Styrene-4-sulfonic acid sodium salt is grafted on poly(ethersulfone) membrane to determine the fractionation of proteins with similar molecular weights. It is used as sodium based ion exchange membranes for membrane capacitive deionization (MCDI) for synthesis. It is used in the study of copolymerization characteristics of sodium styrenesulfonate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2695-37-6
Molecular Formula C8H7NaO3S
Molecular Weight (g/mol) 206.19
MDL Number MFCD00013379 MFCD00084449
InChI Key XFTALRAZSCGSKN-UHFFFAOYSA-M
Synonym sodium 4-vinylbenzenesulfonate, sodium p-styrenesulfonate, sodium 4-styrenesulfonate, sodium p-styrene sulfonate, unii-0kp0v3og5g, benzenesulfonic acid, 4-ethenyl-, sodium salt, poly sodium 4-vinylbenzenesulfonate, 0kp0v3og5g, sodium 4-ethenylbenzenesulfonate, p-styrenesulfonic acid sodium salt
PubChem CID 3571582
IUPAC Name sodium;4-ethenylbenzenesulfonate
SMILES [Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1

Specifications

Color White
Quantity 50 g
Beilstein 3575931
Solubility Information Soluble in water.
Formula Weight 206.19
Physical Form Powder
Chemical Name or Material Styrene-4-sulfonic acid sodium salt
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 220-266-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.