Learn More
Sigma Organic Chemistry Poly(styrene-alt-maleic acid) sodium salt solution | 250ML | 25736-61-2 | MFCD00217732
Supplier: Sigma Organic Chemistry 662631250ML
Poly(styrene-alt-maleic acid) sodium salt solution, 250ML
About This Item:
Density: 1.070 g/mL at 25 C
Refractive Index: n20/D 1.362
Storage: room temp
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.