Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SU 3327
GREENER_CHOICE

Catalog No. p-5546047
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-071-0R 10 mg
36-075-0R 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 36-071-0R Supplier Tocris Bioscience™ Supplier No. 3607/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective JNK inhibitor

SU 3327 is a selective inhibitor of c-Jun N-terminal kinase (JNK) (IC50 = 0.7 μM). Inhibits the protein-protein interaction between JNK and JIP (IC50 = 239 nM). Displays selectivity over p38 MAPK and Akt. Restores insulin sensitivity in a mouse model of type-2 diabetes. Displays antibiotic activity against a range of bacteria including S.aureus, A.baumanni, C.difficile and M.tuberculosis.

Specifications

Quantity 10 mg
Product Type SU 3327
Purity >99%
Molecular Weight (g/mol) 261.3
Inhibitors SU 3327

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.