Learn More
Sigma Aldrich Fine Chemicals Biosciences Monoclonal Anti Vimentin a
Supplier:  Sigma Aldrich Fine Chemicals Biosciences V6630100UL
Encompass
Vimentin is encoded by a single-copy gene mapped to human chromosome 10p13. 53kDa vimentin protein belongs to type III intermediate filament (If) family and is specifically expressed in normal mesenchymal cells. Vimentin protein with 466 amino acids is characterized with a highly conserved a-helical rod domain that is flanked by non -helical amino- terminal head and carboxy-terminal tail domain.
Catalog No. 50-290-5053
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.