Learn More
Sigma Aldrich Fine Chemicals Biosciences Liquid scintillation vials
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1905271PAK
Encompass
Reliable Wheaton 180 brand low-potassium glass. White 22Amm caps have tops suitable for writing. Vials with caps attached are packed in individual cells in trays of 100.
Catalog No. 50-290-5604
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.