Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Substance P Parameter Assay Kit
GSA_VASDP

Catalog No. SKGE007 Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
6 x 96-well Plates
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
SKGE007 6 x 96-well Plates
KGE007 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. SKGE007 Supplier R&D Systems™ Supplier No. SKGE007
Add to Cart
Add to Cart
This item is not returnable. View return policy

Substance P Parameter Assay Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Substance P Parameter Assay Kit quantifies multi-species Substance P in cell culture supernates (50 ul), serum (50 ul), heparin plasma (50 ul), saliva (50 ul), urine (50 ul).

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Assay Range 39.00 - 2500pg/mL
Assay Sensitivity 43.8pg/mL
Conjugate Biotin
Cross Reactivity Cross-species reactivity not tested. Cross-reactivity observed with 1 or more available related molecules.
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Plasma (Heparin), Saliva, Serum, Urine
Specificity Substance P Levels in Various Samples
Sufficient For 6 x 96-well Plates
Target Species Multi-species
For Use With (Application) ELISA
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (50uL),Heparin Plasma (50uL),Saliva (50uL),Urine (50uL)
Storage Requirements Store the unopened product at -20° to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Synonym neurokinin 1, neurokinin 2, neurokinin A, Neurokinin-1, neuromedin L, neurokinin alpha, neuropeptide K, neuropeptide gamma), neuropeptide gamma, neuropeptide K, NK2, NKA, NKNAneurokinin alpha, NPK, PPT, protachykinin-1, substance K, substance P, TAC1, TAC2neuromedin L, tachykinin 2, tachykinin, precursor 1, tachykinin, precursor 1 (substance K, substance P, neurokinin 1, neurokinin 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.