Learn More
Bachem H-Ala-Ala-Phe-pNA
SDP
Supplier:  Bachem 4002753.0050
Encompass_Preferred
H-Ala-Ala-Phe-pNA, 61043-41-2, C21H25N5O5, Mr 427.46, 50mg. AAF-pNA. Substrate for chymotrypsin and tripeptidyl peptidases I and II.
Catalog No. 50-194-7028