Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinaldehyde Bis(dimethyl Acetal) 98.0+%, TCI America™
SDP

Catalog No. S037125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S037125G 25 g
S03715G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S037125G Supplier TCI America Supplier No. S037125G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6922-39-0
Molecular Formula C8H18O4
Molecular Weight (g/mol) 178.23
MDL Number MFCD00127971
InChI Key BJRGNROMZGSJDC-UHFFFAOYSA-N
Synonym Succinaldehyde Tetramethyl Acetal, 1,1,4,4-Tetramethoxybutane
PubChem CID 11263832
IUPAC Name 1,1,4,4-tetramethoxybutane
SMILES COC(CCC(OC)OC)OC

Specifications

Color Colorless
Boiling Point 89°C
Quantity 25 g
Formula Weight 178.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Succinaldehyde Bis(dimethyl Acetal)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.