Learn More
Bioassay Systems EnzyChrom™ Succinate Assay Kit. For quantitative determination of succinate (succinic acid) in food, beverage, agricultural products, and other biological samples.
SDP
Catalog No. 501078340 Shop All Bioassay Systems Products
Encompass_Preferred
For quantitative determination of succinate (succinic acid) in food, beverage, agricultural products, and other biological samples. Key Features: Fast and sensitive. Use 20 µL sample. Linear detection range 10 to 400 µM for colorimetric assays and 2 to 40 µM for fluorimetric assays. Convenient. The procedure involves adding a single working reagent, and reading the absorbance or fluorescence after 30 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD570nm, FL530/585nm. Samples: succinate in food, beverage, agricultural products, and other biological samples. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: Colorimetric assay: 10 µM; Fluorimetric assay: 2 µM.
Catalog No. 50-107-8340 Supplier Bioassay Systems Supplier No. ESNT100
For quantitative determination of succinate (succinic acid) in food, beverage, agricultural products, and other biological samples. Key Features: Fast and sensitive. Use 20 µL sample. Linear detection range 10 to 400 µM for colorimetric assays and 2 to 40 µM for fluorimetric assays. Convenient. The procedure involves adding a single working reagent, and reading the absorbance or fluorescence after 30 minutes. Room temperature assay. No 37°C heater is needed. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD570nm, FL530/585nm. Samples: succinate in food, beverage, agricultural products, and other biological samples. Species: All. Procedure: Assay takes 30 min. Kit size: 100 tests. Detection limit: Colorimetric assay: 10 µM; Fluorimetric assay: 2 µM.