Learn More
Promega Succinate-Glo JmjC Demethylase/Hydroxylase Assay, 10,000 assays
Catalog No. PRV7991 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10,000 Assays
Catalog No. Quantity
PRV7991 10,000 Assays
1 options
Catalog No. PRV7991 Supplier Promega Supplier No. V7991
Succinate-Glo JmjC Demethylase/Hydroxylase Assay is a bioluminescent assay for detecting the activity of JumonjiC (JmjC) histone demethylases hydroxylases.
  • Sensitive, Bioluminescent Detection of Demethylase Activity
  • Easy to use add−and−read assay format
  • Universal assay can be used with any succinate−producing enzyme
  • Optimized for screening applications; low false hits
Specifications
Quantity 10,000 Assays