Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinimide, 98%

Catalog No. AC348650051
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
5 kg
Packaging:
Glass bottle
Plastic drum
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC348650051 5 kg Plastic drum
AC348650050 5 g Glass bottle
AC348651000 100 g Glass bottle
AC348655000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC348650051 Supplier Thermo Scientific Chemicals Supplier No. 348650051

CAS: 123-56-8 | C4H5NO2 | 99.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-56-8
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.08
MDL Number MFCD00005495
InChI Key KZNICNPSHKQLFF-UHFFFAOYSA-N
Synonym succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
PubChem CID 11439
ChEBI CHEBI:9307
IUPAC Name pyrrolidine-2,5-dione
SMILES C1CC(=O)NC1=O

Specifications

Melting Point 121.0°C to 126.0°C
Color Brown to White
Boiling Point 285.0°C to 290.0°C
Flash Point 201°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic drum
Quantity 5 kg
Beilstein 21,369
Merck Index 15,9001
Solubility Information Solubility in water: 240g/L (20°C). Other solubilities: 1g/24 mL alcohol - 1g/5 mL alcohol (60°C), insoluble in ether and chloroform
Formula Weight 99.08
Percent Purity 98%
Physical Form Powder or Flakes
Chemical Name or Material Succinimide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 204-635-6
RTECSNumber WN2200000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.