Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinonitrile 99.0+%, TCI America™
SDP

Catalog No. S0109100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S0109100G 100 g
S010925G 25 g
S0109500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S0109100G Supplier TCI America Supplier No. S0109100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 110-61-2
Molecular Formula C4H4N2
Molecular Weight (g/mol) 80.09
MDL Number MFCD00001949
InChI Key IAHFWCOBPZCAEA-UHFFFAOYSA-N
Synonym succinonitrile, deprelin, succinodinitrile, ethylene cyanide, dician, dinile, 1,2-dicyanoethane, ethylene dicyanide, disuxyl, succinil
PubChem CID 8062
IUPAC Name butanedinitrile
SMILES N#CCCC#N

Specifications

Color White
Boiling Point 159°C
Quantity 100 g
UN Number 3439
Formula Weight 80.09
Percent Purity ≥99.0% (GC)
Physical Form Crystals
Chemical Name or Material Succinonitrile
EINECSNumber (2)-2771&(2)-1526
RTECSNumber WN3850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.