Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinylcholine chloride, 96%

Catalog No. AC460110250
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460110250 25 g
AC460110050 5 g
AC460111000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC460110250 Supplier Thermo Scientific Chemicals Supplier No. 460110250
Add to Cart

CAS: 71-27-2 | C14H30Cl2N2O4 | 361.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-27-2
Molecular Formula C14H30Cl2N2O4
Molecular Weight (g/mol) 361.3
InChI Key YOEWQQVKRJEPAE-UHFFFAOYSA-L
Synonym succinylcholine chloride, succinyldicholine chloride, suxamethonium chloride, midarine, scoline, succinylcholine dichloride, anectine, lysthenon, succinylforte, lysthenone
PubChem CID 22475
ChEBI CHEBI:61219
IUPAC Name trimethyl-[2-[4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl]oxyethyl]azanium;dichloride
SMILES C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C.[Cl-].[Cl-]

Specifications

Melting Point 160.0°C to 162.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (on dry substance) (Argentometry)
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 361.3
Percent Purity 96%
Chemical Name or Material Succinylcholine chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Precautionary Statement GHS P Statement Wash face, hands and any exposed skin thoroughly after handling. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
EINECSNumber 200-747-4

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.