Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Succinylcholine Chloride, Dihydrate, USP, 96-102%, Spectrum™ Chemical
SDP

Catalog No. 18607061 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-061 500 g Amber Glass Bottle
18-607-062 1 kg Amber Glass Bottle
18-607-059 5 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-061 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S1696500GMBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

S1696, 6101-15-1, C14H30Cl2N2O4·2H2O

Spectrum™ Chemical Succinylcholine Chloride, Dihydrate, USP is a medication used to cause short-term paralysis as part of general anesthesia. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 6101-15-1
Molecular Formula C14H34Cl2N2O6
Molecular Weight (g/mol) 397.33
MDL Number MFCD00150564
InChI Key FFSBEIRFVXGRPR-UHFFFAOYSA-L
IUPAC Name trimethyl[2-({4-oxo-4-[2-(trimethylazaniumyl)ethoxy]butanoyl}oxy)ethyl]azanium dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)OCC[N+](C)(C)C

Specifications

CAS Max % 1
Ignition Residue 0.002
Packaging Amber Glass Bottle
Quantity 500 g
Percent Purity 96 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Succinylcholine Chloride, Dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.